Assay Details
| Gene Symbol: APOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100297.1 | NP_001093767.1 | 2 - 3 | 199 | 145 | |
| GenBank mRNA | BC149962.1 | - | 2 - 3 | 199 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149962.1, NM_001100297.1 transcripts.
Set Membership: |
|

APOL3
Genomic Map