Assay Details
| Gene Symbol: GRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175700.2 | NP_783631.2 | 2 - 3 | 273 | 157 | |
| GenBank mRNA | U95812.1 | - | 2 - 3 | 246 | 157 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRO1| Gene Symbol: GRO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175700.2 | NP_783631.2 | 2 - 3 | 273 | 157 | |
| GenBank mRNA | U95812.1 | - | 2 - 3 | 246 | 157 | |
Set Membership: | 
		   
					
						
							 |