Assay Details
| Gene Symbol: TUBB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034663.2 | NP_001029835.1 | 1 - 2 | 143 | 161 | |
| GenBank mRNA | BC105181.1 | - | 1 - 2 | 145 | 161 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105181.1, NM_001034663.2 transcripts.
Set Membership: |
|

TUBB4B
Genomic Map