Assay Details
Gene Symbol: TUBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037615.2 | NP_001032704.1 | 10 - 11 | 1294 | 74 | |
GenBank mRNA | BC110028.1 | - | 10 - 11 | 1295 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
