Assay Details
Gene Symbol: BOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038518.2 | NP_001033607.1 | 7 - 8 | 1094 | 140 | |
GenBank mRNA | AB008627.1 | - | 7 - 8 | 140 | ||
BC109586.1 | - | 7 - 8 | 1095 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
