Assay Details
| Gene Symbol: MT1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078134.2 | NP_001071602.1 | 1 - 2 | 104 | 147 | |
| GenBank mRNA | BC111615.1 | - | 1 - 2 | 106 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111615.1, NM_001078134.2 transcripts.
Set Membership: |
|

MT1E
Genomic Map