Assay Details
| Gene Symbol: MT1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078134.2 | NP_001071602.1 | 2 - 3 | 166 | 150 | |
| GenBank mRNA | BC111615.1 | - | 2 - 3 | 168 | 150 | |
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the BC111615.1, NM_001078134.2 transcripts.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Mammalian Gene Collection  Bos taurus  Probe spans exons | 
 Back To Top
 Back To Top
 MT1E
MT1E Genomic Map
Genomic Map Back To Top
 Back To Top