Assay Details
| Gene Symbol: GSTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078149.1 | NP_001071617.1 | 5 - 6 | 469 | 154 | |
| GenBank mRNA | BC102540.1 | - | 5 - 6 | 469 | 154 | |
| U49179.1 | - | 5 - 6 | 460 | 154 | ||
More Information
Set Membership: |
|

GSTA1
Genomic Map