Assay Details
| Gene Symbol: CD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046021.1 | NP_001039486.1 | 3 - 4 | 609 | 149 | |
| GenBank mRNA | DQ192542.1 | - | 3 - 4 | 609 | 149 | |
More Information
Set Membership: |
|

CD1B
Genomic Map