Assay Details
| Gene Symbol: BOLA-DQA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013601.3 | NP_001013619.3 | 248 | 133 | ||
| GenBank mRNA | BC118360.1 | - | 248 | 133 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BOLA-DQA1| Gene Symbol: BOLA-DQA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013601.3 | NP_001013619.3 | 248 | 133 | ||
| GenBank mRNA | BC118360.1 | - | 248 | 133 | ||
Set Membership: |
|