Assay Details
| Gene Symbol: SCAMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024566.1 | NP_001019737.1 | 7 | 1955 | 115 | |
| GenBank mRNA | BC113329.1 | - | 7 | 1968 | 115 | |
| BT021641.1 | - | 7 | 1955 | 115 | ||
More Information
Set Membership: |
|

SCAMP4
Genomic Map