Assay Details
| Gene Symbol: RPL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045958.2 | NP_001039423.1 | 1 | 449 | 159 | |
| GenBank mRNA | JX899372.1 | - | 1 | 450 | 159 | |
| Gene Symbol: LOC786430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC114200.1 | - | 1 | 450 | 159 | |
More Information
Set Membership: |
|

RPL23A
Genomic Map