Assay Details
| Gene Symbol: MARCKSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076172.1 | NP_001069640.1 | 2 | 353 | 105 | |
| GenBank mRNA | BC120310.1 | - | 2 | 353 | 105 | |
| BC120422.1 | - | 2 | 328 | 105 | ||
More Information
Set Membership: |
|

MARCKSL1
Genomic Map