Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076479.1 | NP_001069947.1 | 4 | 556 | 85 | |
| GenBank mRNA | BC112524.1 | - | 4 | 556 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNRPD3| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076479.1 | NP_001069947.1 | 4 | 556 | 85 | |
| GenBank mRNA | BC112524.1 | - | 4 | 556 | 85 | |
Set Membership: |
|