Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077955.1 | NP_001071423.1 | 3 | 1147 | 105 | |
| GenBank mRNA | BC103342.1 | - | 3 | 1147 | 105 | |
More Information
Set Membership: |
|

ZNF394
Genomic Map