Assay Details
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077957.1 | NP_001071425.1 | 5 | 1451 | 139 | |
GenBank mRNA | BC123500.1 | - | 5 | 1451 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
