Assay Details
Gene Symbol: RPL36AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079508.2 | NP_001072976.1 | 51 | 69 | ||
GenBank mRNA | BC102168.1 | - | 53 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102168.1, NM_001079508.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
