Assay Details
| Gene Symbol: VTI1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079792.2 | NP_001073260.1 | 4 | 1750 | 130 | |
| GenBank mRNA | BC126647.1 | - | 4 | 1751 | 130 | |
More Information
Set Membership: |
|

VTI1A
Genomic Map