Assay Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080912.1 | NP_001074381.1 | 7 | 1552 | 119 | |
| GenBank mRNA | BC119882.1 | - | 7 | 1552 | 119 | |
More Information
Set Membership: |
|

LHPP
Genomic Map