Assay Details
| Gene Symbol: FZD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101048.1 | NP_001094518.1 | 1 | 3849 | 130 | |
| GenBank mRNA | BC151283.1 | - | 1 | 3849 | 130 | |
More Information
Set Membership: |
|

FZD1
Genomic Map