Assay Details
| Gene Symbol: XRCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102354.1 | NP_001095824.1 | 3 | 2650 | 112 | |
| GenBank mRNA | BC151285.1 | - | 3 | 2650 | 112 | |
More Information
Set Membership: |
|

XRCC2
Genomic Map