Assay Details
| Gene Symbol: JUND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103253.1 | NP_001096723.1 | 1 | 1655 | 146 | |
| GenBank mRNA | BC151336.1 | - | 1 | 1655 | 146 | |
More Information
Set Membership: |
|

JUND
Genomic Map