Assay Details
Gene Symbol: GALNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103321.1 | NP_001096791.1 | 1 | 4446 | 78 | |
GenBank mRNA | BC151593.1 | - | 1 | 4446 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
