Assay Details
| Gene Symbol: MSRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174114.2 | NP_776539.1 | 6 | 750 | 155 | |
| GenBank mRNA | BC102980.1 | - | 6 | 747 | 155 | |
| BT021044.1 | - | 6 | 735 | 155 | ||
| U37150.1 | - | 6 | 750 | 155 | ||
More Information
Set Membership: |
|

MSRA
Genomic Map