Assay Details
| Gene Symbol: CXCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174360.3 | NP_776785.2 | 2 | 1713 | 122 | |
| GenBank mRNA | U19947.1 | - | 1 | 1648 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCR1| Gene Symbol: CXCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174360.3 | NP_776785.2 | 2 | 1713 | 122 | |
| GenBank mRNA | U19947.1 | - | 1 | 1648 | 122 | |
Set Membership: |
|