Assay Details
| Gene Symbol: UQCRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174805.2 | NP_777230.1 | 2 | 445 | 115 | |
| GenBank mRNA | L06665.1 | - | 2 | 445 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UQCRQ| Gene Symbol: UQCRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174805.2 | NP_777230.1 | 2 | 445 | 115 | |
| GenBank mRNA | L06665.1 | - | 2 | 445 | 115 | |
Set Membership: |
|