Assay Details
Gene Symbol: AHSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177522.2 | NP_803488.1 | 3 | 355 | 119 | |
GenBank mRNA | AF485326.1 | - | 3 | 355 | 119 | |
BC103281.1 | - | 3 | 362 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
