Assay Details
| Gene Symbol: MRPL43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174563.3 | NP_776988.2 | 3 | 579 | 140 | |
| GenBank mRNA | BC112678.1 | - | 3 | 580 | 140 | |
| CN441629.1 | - | 1 | 302 | 140 | ||
| M34915.1 | - | 446 | 142 | |||
More Information
Set Membership: |
|

MRPL43
Genomic Map