Assay Details
Gene Symbol: LAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203435.4 | NP_982259.3 | 2 | 214 | 121 | |
GenBank mRNA | AY911374.1 | - | 2 | 216 | 121 | |
S76279.1 | - | 2 | 154 | 121 | ||
U48357.1 | - | 2 | 127 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
