Assay Details
| Gene Symbol: IFITM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078054.2 | NP_001071522.1 | 1 | 45 | 94 | |
| GenBank mRNA | BC102339.1 | - | 1 | 46 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102339.1, NM_001078054.2 transcripts.
Set Membership: |
|

IFITM2
Genomic Map