Assay Details
Gene Symbol: PGRMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099060.1 | NP_001092530.1 | 3 | 1745 | 144 | |
GenBank mRNA | BC142203.1 | - | 3 | 1745 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
