Assay Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038147.1 | 3 - 4 | 842 | 67 | ||
| NR_038148.1 | 2 - 3 | 797 | 67 | |||
| GenBank mRNA | EH132547.1 | - | 2 - 3 | 406 | 67 | |
| EU848421.1 | - | 3 - 4 | 843 | 67 | ||
More Information
Set Membership: |
|

SNHG12
Genomic Map