Assay Details
| Gene Symbol: TMEM139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143872.1 | NP_001137344.1 | 1 - 2 | 199 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143872.1 transcripts.
Set Membership: |
|

TMEM139
Genomic Map