Assay Details
| Gene Symbol: CRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144097.1 | NP_001137569.1 | 1 - 2 | 164 | 68 | |
| GenBank mRNA | AB255049.1 | - | 1 - 2 | 104 | 68 | |
More Information
Set Membership: |
|

CRP
Genomic Map