Assay Details
| Gene Symbol: ASAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075459.2 | NP_001068927.1 | 8 - 9 | 726 | 64 | |
| GenBank mRNA | BC118455.1 | - | 8 - 9 | 727 | 64 | |
More Information
Set Membership: |
|

ASAH1
Genomic Map