Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105611.2 | NP_001099081.1 | 6 - 7 | 599 | 76 | |
| GenBank mRNA | BC134765.1 | - | 6 - 7 | 600 | 76 | |
| U57745.1 | - | 6 - 7 | 559 | 76 | ||
More Information
Set Membership: |
|

CD40
Genomic Map