Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127440.1 | NP_001120912.1 | 1 - 2 | 1070 | 103 | |
| GenBank mRNA | AJ936934.2 | - | 1 - 2 | 1070 | 103 | |
More Information
Set Membership: |
|

ZAR1L
Genomic Map