Assay Details
Gene Symbol: TLE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128497.2 | NP_001121969.1 | 4 - 5 | 338 | 73 | |
GenBank mRNA | BC103327.1 | - | 4 - 5 | 340 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
