Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128504.2 | NP_001121976.1 | 2 - 3 | 268 | 65 | |
| GenBank mRNA | BC133471.1 | - | 2 - 3 | 269 | 65 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map