Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128504.2 | NP_001121976.1 | 4 - 5 | 560 | 69 | |
| GenBank mRNA | BC133471.1 | - | 4 - 5 | 561 | 69 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map