Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128505.2 | NP_001121977.1 | 5 - 6 | 630 | 72 | |
| GenBank mRNA | BC133324.2 | - | 5 - 6 | 630 | 72 | |
More Information
Set Membership: |
|

MBD3
Genomic Map