Assay Details
| Gene Symbol: FGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078011.2 | NP_001071479.2 | 2 - 3 | 471 | 98 | |
| GenBank mRNA | EF042192.1 | - | 2 - 3 | 471 | 98 | |
More Information
Set Membership: |
|

FGF5
Genomic Map