Assay Details
| Gene Symbol: CRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079649.1 | NP_001073117.1 | 7 - 8 | 570 | 79 | |
| GenBank mRNA | BC118323.1 | - | 7 - 8 | 570 | 79 | |
More Information
Set Membership: |
|

CRIP2
Genomic Map