Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099163.1 | NP_001092633.1 | 2 - 3 | 272 | 128 | |
| GenBank mRNA | BC142155.1 | - | 2 - 3 | 272 | 128 | |
More Information
Set Membership: |
|

PXMP4
Genomic Map