Assay Details
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099950.1 | NP_001093420.1 | 2 - 3 | 588 | 85 | |
GenBank mRNA | BC133508.1 | - | 2 - 3 | 588 | 85 | |
BT030589.2 | - | 2 - 3 | 533 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
