Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101985.1 | NP_001095455.1 | 5 - 6 | 833 | 62 | |
| GenBank mRNA | BC149096.1 | - | 5 - 6 | 833 | 62 | |
More Information
Set Membership: |
|

FBXW4
Genomic Map