Assay Details
| Gene Symbol: RETSAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102279.2 | NP_001095749.2 | 1 - 2 | 210 | 71 | |
| GenBank mRNA | BC149535.1 | - | 1 - 2 | 217 | 71 | |
More Information
Set Membership: |
|

RETSAT
Genomic Map