Assay Details
| Gene Symbol: AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105482.1 | NP_001098952.1 | 5 - 6 | 796 | 58 | |
| GenBank mRNA | BC122671.1 | - | 5 - 6 | 796 | 58 | |
More Information
Set Membership: |
|

AARSD1
Genomic Map