Assay Details
| Gene Symbol: SELENOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163225.1 | NP_001156697.1 | 4 - 5 | 482 | 57 | |
| GenBank mRNA | BC103044.1 | - | 4 - 5 | 257 | 57 | |
More Information
Set Membership: |
|

SELENOW
Genomic Map