Assay Details
| Gene Symbol: VNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163920.1 | NP_001157392.1 | 5 - 6 | 1199 | 92 | |
| GenBank mRNA | BT030550.1 | - | 5 - 6 | 1199 | 92 | |
More Information
Set Membership: |
|

VNN2
Genomic Map